Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533345_m1
          See other FAM186B GE Assays ›
          Gene Symbol:
          Fam186b
          Gene Name:
          family with sequence similarity 186, member B
          Gene Aliases:
          RGD1559522
          Chromosome Location:
          Chr.7: 140935645 - 140951071 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533345_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam186b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134633.1 NP_001128105.1 3 - 4 636 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003878.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P106042 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P132862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921335 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 745043 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157192 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132862 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P106042 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P132862 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P132862 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P106042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10907119 - -
          Affymetrix Rat Gene 1.0 ST Array 10907112 - -
          Affymetrix Rat Gene 1.0 ST Array 10907117 - -
          Affymetrix Rat Gene 1.0 ST Array 10907121 - -
          Affymetrix Rat Gene 1.0 ST Array 10907114 - -
          Affymetrix Rat Gene 1.0 ST Array 10907116 - -
          Affymetrix Rat Gene 1.0 ST Array 10907113 - -
          Affymetrix Rat Gene 1.0 ST Array 10907115 - -
          Affymetrix Rat Gene 1.0 ST Array 10907120 - -
          Affymetrix Rat Gene 1.0 ST Array 10907118 - -
          Affymetrix Rat Gene 1.1 ST Array 10907111 - -
          Affymetrix Rat Clariom S Assay PSR0700031266.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031264.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031268.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031262.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031263.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031265.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700031267.rn.1 TC0700003878.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10907111 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21477438 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369251 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline