Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533624_m1
          See other GPR89B GE Assays ›
          Gene Symbol:
          Gpr89b
          Gene Name:
          G protein-coupled receptor 89B
          Gene Aliases:
          Gpr89, RGD1304837
          Chromosome Location:
          Chr.2: 198999945 - 199038702 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533624_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpr89b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001139486.1 NP_001132958.1 12 - 13 1190 116
          XM_006232976.2 XP_006233038.1 10 - 11 1179 116
          GenBank mRNA AY724489.1 - 12 - 13 1121 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004267.rn.1

          Set Membership:

          Amplicon length greater than or equal to 101 Protein Kinase 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          skin development
          regulation of anion transport
          intracellular pH reduction
          signal transducer activity
          voltage-gated anion channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P386936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569975 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3141 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152624 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119198 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386936 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P386936 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P386936 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P386936 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825224 - -
          Affymetrix Rat Gene 1.0 ST Array 10825222 - -
          Affymetrix Rat Gene 1.0 ST Array 10825214 - -
          Affymetrix Rat Gene 1.0 ST Array 10825220 - -
          Affymetrix Rat Gene 1.0 ST Array 10825218 - -
          Affymetrix Rat Gene 1.0 ST Array 10825221 - -
          Affymetrix Rat Gene 1.0 ST Array 10825215 - -
          Affymetrix Rat Gene 1.0 ST Array 10825225 - -
          Affymetrix Rat Gene 1.0 ST Array 10825216 - -
          Affymetrix Rat Gene 1.0 ST Array 10825223 - -
          Affymetrix Rat Gene 1.0 ST Array 10825217 - -
          Affymetrix Rat Gene 1.0 ST Array 10825219 - -
          Affymetrix Rat Gene 1.1 ST Array 10825213 - -
          Rat Genome 230 2.0 Array 1372313_at - -
          Rat Genome 230 2.0 Array 1372313_PM_at - -
          Affymetrix HT Rat Focus Array 1372313_at - -
          Affymetrix HT Rat Focus Array 1372313_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200029304.rn.1 TC0200004267.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029303.rn.1 TC0200004267.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029305.rn.1 TC0200004267.rn.1 RTA TC
          Rat Expression Set 230 1372313_at - -
          Rat Expression Set 230 1372313_PM_at - -
          Rat Genome U34 Set rc_AI070732_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21081929 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1374215 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365884 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0