Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01533986_m1
          See other ATP1A1 GE Assays ›
          Gene Symbol:
          Atp1a1
          Gene Name:
          ATPase, Na+/K+ transporting, alpha 1 polypeptide
          Gene Aliases:
          Nkaa1b
          Chromosome Location:
          Chr.2: 204003742 - 204032023 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01533986_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012504.1 NP_036636.1 14 - 15 2202 72
          GenBank mRNA BC061968.1 - 14 - 15 2213 5598014 72
          M14511.1 - 14 - 15 2202 72
          M28647.1 - 14 - 15 2225 72
          X05882.1 - 14 - 15 1979 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004336.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Transporter Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1a1

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of the force of heart contraction
          regulation of sodium ion transport
          potassium ion transport
          sodium ion transport
          cellular sodium ion homeostasis
          regulation of blood pressure
          potassium ion import
          energy coupled proton transmembrane transport, against electrochemical gradient
          dephosphorylation
          cellular potassium ion homeostasis
          negative regulation of glucocorticoid biosynthetic process
          sodium ion transmembrane transport
          sodium ion export from cell
          response to drug
          negative regulation of heart contraction
          positive regulation of heart contraction
          positive regulation of striated muscle contraction
          relaxation of cardiac muscle
          cardiac muscle contraction
          membrane hyperpolarization
          cellular response to mechanical stimulus
          cellular response to steroid hormone stimulus
          potassium ion transmembrane transport
          regulation of cardiac muscle cell contraction
          membrane repolarization
          response to glycoside
          sodium:potassium-exchanging ATPase activity
          protein binding
          ATP binding
          phosphatase activity
          protein kinase binding
          protein domain specific binding
          ankyrin binding
          potassium ion binding
          sodium ion binding
          ADP binding
          phosphatidylinositol 3-kinase binding
          chaperone binding
          steroid hormone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P783572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P783572 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P783572 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P783572 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P184279 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P184273 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700494491 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 702033580 - -
          Agilent Rat cDNA Microarray (G4105A) 700494491 - -
          Agilent Rat cDNA Microarray (G4105A) 702033580 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10825486 - -
          Affymetrix Rat Gene 1.0 ST Array 10825485 - -
          Affymetrix Rat Gene 1.0 ST Array 10825478 - -
          Affymetrix Rat Gene 1.0 ST Array 10825481 - -
          Affymetrix Rat Gene 1.0 ST Array 10825479 - -
          Affymetrix Rat Gene 1.0 ST Array 10825476 - -
          Affymetrix Rat Gene 1.0 ST Array 10825484 - -
          Affymetrix Rat Gene 1.0 ST Array 10825475 - -
          Affymetrix Rat Gene 1.0 ST Array 10825483 - -
          Affymetrix Rat Gene 1.0 ST Array 10825473 - -
          Affymetrix Rat Gene 1.0 ST Array 10825474 - -
          Affymetrix Rat Gene 1.0 ST Array 10825480 - -
          Affymetrix Rat Gene 1.0 ST Array 10825482 - -
          Affymetrix Rat Gene 1.0 ST Array 10825477 - -
          Affymetrix Rat Gene 1.0 ST Array 10825487 - -
          Affymetrix Rat Gene 1.1 ST Array 10825472 - -
          Rat Genome 230 2.0 Array 1367585_a_at - -
          Rat Genome 230 2.0 Array 1367585_PM_a_at - -
          Rat Genome 230 2.0 Array 1371108_a_at - -
          Rat Genome 230 2.0 Array 1371108_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371108_PM_a_at - -
          Affymetrix HT Rat Focus Array 1371108_a_at - -
          Affymetrix HT Rat Focus Array 1367585_PM_a_at - -
          Affymetrix HT Rat Focus Array 1367585_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200029833.rn.1 TC0200004336.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200029834.rn.1 TC0200004336.rn.1 RTA TC
          Rat Expression Set 230 1367585_a_at - -
          Rat Expression Set 230 1367585_PM_a_at - -
          Rat Expression Set 230 1371108_PM_a_at - -
          Rat Expression Set 230 1371108_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10825472 - -
          Rat Genome U34 Set M28647_g_at - -
          Rat Genome U34 Set M74494_at - -
          Rat Genome U34 Set M28647_at - -
          Rat Genome U34 Set M74494_g_at - -
          Rat Genome U34 Set rc_AI072060_s_at - -
          Rat Genome U34 Set M74494_g_at - -
          Rat Genome U34 Set rc_AI072060_s_at - -
          Rat Genome U34 Set M74494_at - -
          Rat Genome U34 Set M28647_g_at - -
          Rat Genome U34 Set M28647_at - -
          Rat Genome U34 Set M28647_at - -
          Rat Genome U34 Set M28647_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22186749 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351569 - -
          RatRef-12 Expression BeadChip ILMN_57453 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline