Search
Search
| Gene Symbol: Tdrkh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014038.2 | NP_001014060.2 | 3 - 4 | 345 | 68 | |
| XM_006232870.2 | XP_006232932.1 | 3 - 4 | 348 | 68 | ||
| XM_006232871.2 | XP_006232933.1 | 3 - 4 | 2304 | 68 | ||
| GenBank mRNA | BC059161.1 | - | 3 - 4 | 301 | 6887770 | 68 |
Set Membership: |
|