Search
Search
| Gene Symbol: Gdap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013201.1 | NP_001013219.1 | 11 - 12 | 1465 | 88 | |
| XM_006233032.2 | XP_006233094.1 | 12 - 13 | 1563 | 88 | ||
| XM_006233033.2 | XP_006233095.1 | 11 - 12 | 1513 | 88 | ||
| GenBank mRNA | BC082000.1 | - | 11 - 12 | 1465 | 7131173 | 88 |
| FQ211326.1 | - | 13 - 14 | 1646 | 88 | ||
Set Membership: |
|