Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01534636_m1
          See other KPNA3 GE Assays ›
          Gene Symbol:
          Kpna3
          Gene Name:
          karyopherin alpha 3
          Gene Aliases:
          importin
          Chromosome Location:
          Chr.15: 41696356 - 41770112 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01534636_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kpna3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001014792.1 NP_001014792.1 10 - 11 771 65
          XM_006252127.2 XP_006252189.1 10 - 11 865 65
          XM_008770761.1 XP_008768983.1 10 - 11 720 65
          GenBank mRNA AY779027.1 - 10 - 11 771 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500001839.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kpna3

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          protein C-terminus binding
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P238803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 453565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 111328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310862 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24296 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 280769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 440312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 666075 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140056 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163242 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152239 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112243 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163907 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 110905 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50639 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150463 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39375 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77288 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238803 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P238803 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P238803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P238803 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P355152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83751 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94475 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94720 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10784369 - -
          Affymetrix Rat Gene 1.0 ST Array 10784368 - -
          Affymetrix Rat Gene 1.0 ST Array 10784377 - -
          Affymetrix Rat Gene 1.0 ST Array 10784375 - -
          Affymetrix Rat Gene 1.0 ST Array 10784376 - -
          Affymetrix Rat Gene 1.0 ST Array 10784364 - -
          Affymetrix Rat Gene 1.0 ST Array 10784362 - -
          Affymetrix Rat Gene 1.0 ST Array 10784367 - -
          Affymetrix Rat Gene 1.0 ST Array 10784363 - -
          Affymetrix Rat Gene 1.0 ST Array 10784374 - -
          Affymetrix Rat Gene 1.0 ST Array 10784365 - -
          Affymetrix Rat Gene 1.0 ST Array 10784372 - -
          Affymetrix Rat Gene 1.0 ST Array 10784370 - -
          Affymetrix Rat Gene 1.0 ST Array 10784371 - -
          Affymetrix Rat Gene 1.0 ST Array 10784373 - -
          Affymetrix Rat Gene 1.0 ST Array 10784366 - -
          Affymetrix Rat Gene 1.0 ST Array 10784361 - -
          Affymetrix Rat Gene 1.1 ST Array 10784360 - -
          Rat Genome 230 2.0 Array 1373116_PM_at - -
          Rat Genome 230 2.0 Array 1373116_at - -
          Affymetrix HT Rat Focus Array 1373116_at - -
          Affymetrix HT Rat Focus Array 1373116_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500012326.rn.1 TC1500001839.rn.1 RTA TC
          Rat Expression Set 230 1373116_at - -
          Rat Expression Set 230 1373116_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10784360 - -
          Rat Genome U34 Set rc_AI011082_at - -
          Rat Genome U34 Set rc_AA945320_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21530498 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366672 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline