Search
Search
| Gene Symbol: Fbxo16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013132.2 | NP_001013150.1 | 2 - 3 | 263 | 65 | |
| XM_006252201.2 | XP_006252263.1 | 3 - 4 | 1036 | 65 | ||
| XM_006252202.2 | XP_006252264.1 | 2 - 3 | 916 | 65 | ||
| GenBank mRNA | BC062801.1 | - | 2 - 3 | 266 | 6889991 | 65 |
| FQ213407.1 | - | 2 - 3 | 197 | 65 | ||
Set Membership: |
|