Search
Search

Adamtsl5| Gene Symbol: Adamtsl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108071.1 | NP_001101541.1 | 3 - 4 | 351 | 98 | |
| XM_006240915.2 | XP_006240977.1 | 5 - 6 | 1280 | 98 | ||
| XM_006240916.2 | XP_006240978.1 | 5 - 6 | 1270 | 98 | ||
| XM_006240917.2 | XP_006240979.1 | 5 - 6 | 1277 | 98 | ||
| XM_006240918.2 | XP_006240980.1 | 5 - 6 | 1220 | 98 | ||
| XM_006240919.2 | XP_006240981.1 | 5 - 6 | 1279 | 98 | ||
| XM_006240920.2 | XP_006240982.1 | 4 - 5 | 1109 | 98 | ||
| GenBank mRNA | FQ224249.1 | - | 5 - 6 | 580 | 98 | |
Set Membership: |
|