Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01535451_m1
          See other RNF126 GE Assays ›
          Gene Symbol:
          Rnf126
          Gene Name:
          ring finger protein 126
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 12820812 - 12828325 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01535451_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf126
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033702.1 NP_001028874.1 1 - 2 137 84
          XM_006240899.1 XP_006240961.1 1 - 2 132 84
          GenBank mRNA BC099810.1 - 1 - 2 137 5621533 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000230.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf126

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein monoubiquitination
          regulation of protein localization
          negative regulation of epidermal growth factor receptor signaling pathway
          regulation of cell proliferation
          retrograde transport, endosome to Golgi
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          protein K63-linked ubiquitination
          protein K48-linked ubiquitination
          cytoplasm-associated proteasomal ubiquitin-dependent protein catabolic process
          epidermal growth factor receptor binding
          zinc ion binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P083134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319138 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 492033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268845 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083134 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P083134 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P083134 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P536985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48699 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10894022 - -
          Affymetrix Rat Gene 1.0 ST Array 10894025 - -
          Affymetrix Rat Gene 1.0 ST Array 10894019 - -
          Affymetrix Rat Gene 1.0 ST Array 10894021 - -
          Affymetrix Rat Gene 1.0 ST Array 10894023 - -
          Affymetrix Rat Gene 1.0 ST Array 10894018 - -
          Affymetrix Rat Gene 1.0 ST Array 10894020 - -
          Affymetrix Rat Gene 1.0 ST Array 10894027 - -
          Affymetrix Rat Gene 1.0 ST Array 10894026 - -
          Affymetrix Rat Gene 1.0 ST Array 10894024 - -
          Affymetrix Rat Gene 1.1 ST Array 10894017 - -
          Rat Genome 230 2.0 Array 1381764_PM_s_at - -
          Rat Genome 230 2.0 Array 1376639_PM_at - -
          Rat Genome 230 2.0 Array 1381764_s_at - -
          Rat Genome 230 2.0 Array 1376639_at - -
          Affymetrix HT Rat Focus Array 1381764_s_at - -
          Affymetrix HT Rat Focus Array 1376639_at - -
          Affymetrix HT Rat Focus Array 1381764_PM_s_at - -
          Affymetrix HT Rat Focus Array 1376639_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700002186.rn.1 TC0700000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002187.rn.1 TC0700000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002188.rn.1 TC0700000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002190.rn.1 TC0700000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002191.rn.1 TC0700000230.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700002193.rn.1 TC0700000230.rn.1 RTA TC
          Rat Expression Set 230 1376639_at - -
          Rat Expression Set 230 1376639_PM_at - -
          Rat Expression Set 230 1381764_PM_s_at - -
          Rat Expression Set 230 1381764_s_at - -
          Affymetrix Rat Gene 1.0 ST Array 10894017 - -
          Rat Genome U34 Set rc_AA819792_at - -
          Rat Genome U34 Set rc_AI230424_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20728085 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline