Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01535747_g1
          See other EIF6 GE Assays ›
          Gene Symbol:
          Eif6
          Gene Name:
          eukaryotic translation initiation factor 6
          Gene Aliases:
          Itgb4bp, eIF-6
          Chromosome Location:
          Chr.3: 151356867 - 151363188 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01535747_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037352.1 NP_001032429.1 1 - 2 327 104
          XM_006235316.2 XP_006235378.1 2 - 3 220 104
          GenBank mRNA BC105764.1 - 1 - 2 327 7929717 104
          FQ220694.1 - 1 - 2 103 104
          FQ220773.1 - 2 - 3 177 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300004558.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif6

          Panther Classification:

          Molecular Function -

          translation factor translation initiation factor translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal subunit export from nucleus
          translational initiation
          mature ribosome assembly
          ribosomal large subunit biogenesis
          translation initiation factor activity
          ribosome binding
          ribosomal large subunit binding

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118614 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118614 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118614 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1005770 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10778387 - -
          Affymetrix Rat Gene 1.0 ST Array 10778389 - -
          Affymetrix Rat Gene 1.0 ST Array 10778388 - -
          Affymetrix Rat Gene 1.1 ST Array 10778386 - -
          Rat Genome 230 2.0 Array 1388507_at - -
          Rat Genome 230 2.0 Array 1388507_PM_at - -
          Affymetrix HT Rat Focus Array 1388507_PM_at - -
          Affymetrix HT Rat Focus Array 1388507_at - -
          Affymetrix Rat Clariom S Assay PSR0300034973.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034975.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034970.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034969.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034971.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034968.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034972.rn.1 TC0300004558.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300034974.rn.1 TC0300004558.rn.1 RTA TC
          Rat Expression Set 230 1388507_PM_at - -
          Rat Expression Set 230 1388507_at - -
          Affymetrix Rat Gene 1.0 ST Array 10778386 - -
          Rat Genome U34 Set rc_AA998690_f_at - -
          Rat Genome U34 Set rc_AI104279_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-5xfkf:80/100.66.78.86:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline