Search
Search

RGD1561517| Gene Symbol: RGD1561517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135901.1 | NP_001129373.1 | 2 - 3 | 244 | 69 | |
| XM_006235356.2 | XP_006235418.1 | 2 - 3 | 1652 | 69 | ||
| GenBank mRNA | DQ080432.1 | - | 2 - 3 | 244 | 69 | |
Set Membership: |
|