Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01535871_m1
          See other RALY GE Assays ›
          Gene Symbol:
          Raly
          Gene Name:
          RALY heterogeneous nuclear ribonucleoprotein
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 150322834 - 150387345 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01535871_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Raly
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011958.1 NP_001011958.1 8 - 9 1051 72
          XM_008762314.1 XP_008760536.1 9 - 10 1489 72
          XM_008762315.1 XP_008760537.1 8 - 9 1207 72
          XM_008762316.1 XP_008760538.1 8 - 9 1119 72
          XM_008762317.1 XP_008760539.1 7 - 8 1155 72
          GenBank mRNA BC087074.1 - 8 - 9 1051 7134486 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002106.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Raly

          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P363597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 192065 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 937861 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 933575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 638859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106055 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 62723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80294 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54644 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13912 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26454 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363597 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P363597 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P363597 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1043535 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P363597 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6304 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88239 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92820 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10841292 - -
          Affymetrix Rat Gene 1.0 ST Array 10841293 - -
          Affymetrix Rat Gene 1.0 ST Array 10841291 - -
          Affymetrix Rat Gene 1.0 ST Array 10841288 - -
          Affymetrix Rat Gene 1.0 ST Array 10841289 - -
          Affymetrix Rat Gene 1.1 ST Array 10841287 - -
          Rat Genome 230 2.0 Array 1388363_PM_at - -
          Rat Genome 230 2.0 Array 1388363_at - -
          Affymetrix HT Rat Focus Array 1388363_PM_at - -
          Affymetrix HT Rat Focus Array 1388363_at - -
          Affymetrix Rat Clariom S Assay PSR0300015394.rn.1 TC0300002106.rn.1 RTA TC
          Rat Expression Set 230 1388363_at - -
          Rat Expression Set 230 1388363_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10841287 - -
          Rat Genome U34 Set rc_AA965238_at - -
          Rat Genome U34 Set rc_AI071688_at - -
          Rat Genome U34 Set rc_AA818261_s_at - -
          Rat Genome U34 Set rc_AI169211_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20722041 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline