Search
Search
| Gene Symbol: Snta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100901.1 | NP_001094371.1 | 3 - 4 | 790 | 84 | |
| XM_006235326.2 | XP_006235388.1 | 3 - 4 | 829 | 84 | ||
| GenBank mRNA | BC100086.1 | - | 1 - 2 | 86 | 84 | |
| BC169101.1 | - | 3 - 4 | 790 | 5599966 | 84 | |
Set Membership: |
|