Search
Search
| Gene Symbol: Gins1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109207.2 | NP_001102677.2 | 3 - 4 | 349 | 63 | |
| GenBank mRNA | BC166989.1 | - | 3 - 4 | 349 | 9098732 | 63 |
| FQ222087.1 | - | 1 - 2 | 103 | 63 | ||
Set Membership: |
|