Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536463_g1
          See other SLC52A3 GE Assays ›
          Gene Symbol:
          Slc52a3
          Gene Name:
          solute carrier family 52, riboflavin transporter, member 3
          Gene Aliases:
          RGD1304644, Rft2, rRFT2
          Chromosome Location:
          Chr.3: 147503770 - 147516475 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536463_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc52a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001037198.1 NP_001032275.1 3 - 4 1202 73
          XM_006235238.2 XP_006235300.1 6 - 7 1873 73
          XM_008762327.1 XP_008760549.1 6 - 7 2253 73
          XM_008762328.1 XP_008760550.1 3 - 4 2064 73
          XM_008762329.1 XP_008760551.1 2 - 3 1577 73
          GenBank mRNA BC099097.1 - 3 - 4 1202 7459579 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300002020.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc52a3

          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          sensory perception of sound
          riboflavin transport
          cellular response to heat
          riboflavin transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P838822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252280 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P838822 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P838822 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P838822 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P309420 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10840786 - -
          Affymetrix Rat Gene 1.0 ST Array 10840785 - -
          Affymetrix Rat Gene 1.0 ST Array 10840787 - -
          Affymetrix Rat Gene 1.0 ST Array 10840784 - -
          Affymetrix Rat Gene 1.1 ST Array 10840783 - -
          Rat Genome 230 2.0 Array 1377088_PM_at - -
          Rat Genome 230 2.0 Array 1377088_at - -
          Affymetrix HT Rat Focus Array 1377088_at - -
          Affymetrix HT Rat Focus Array 1377088_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300014397.rn.1 TC0300002020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014396.rn.1 TC0300002020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014399.rn.1 TC0300002020.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300014400.rn.1 TC0300002020.rn.1 RTA TC
          Rat Expression Set 230 1377088_at - -
          Rat Expression Set 230 1377088_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10840783 - -
          Rat Genome U34 Set rc_AA891844_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21149595 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350396 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline