Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536526_g1
          See other FAM96B GE Assays ›
          Gene Symbol:
          Fam96b
          Gene Name:
          family with sequence similarity 96, member B
          Gene Aliases:
          RGD1311762
          Chromosome Location:
          Chr.19: 551708 - 553573 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536526_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam96b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001144854.1 NP_001138326.1 4 - 5 442 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000013.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          chromosome segregation
          biological_process
          iron-sulfur cluster assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P102870 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P007921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102870 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007921 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P007921 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P102870 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P102870 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P007921 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10805643 - -
          Affymetrix Rat Gene 1.0 ST Array 10805641 - -
          Affymetrix Rat Gene 1.0 ST Array 10805642 - -
          Affymetrix Rat Gene 1.0 ST Array 10805645 - -
          Affymetrix Rat Gene 1.0 ST Array 10805644 - -
          Affymetrix Rat Gene 1.1 ST Array 10805640 - -
          Rat Genome 230 2.0 Array 1372001_at - -
          Rat Genome 230 2.0 Array 1372001_PM_at - -
          Affymetrix HT Rat Focus Array 1372001_at - -
          Affymetrix HT Rat Focus Array 1372001_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900000119.rn.1 TC1900000013.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900000117.rn.1 TC1900000013.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900000118.rn.1 TC1900000013.rn.1 RTA TC
          Rat Expression Set 230 1372001_PM_at - -
          Rat Expression Set 230 1372001_at - -
          Rat Genome U34 Set rc_AA892010_at - -
          Rat Genome U34 Set rc_AA892010_g_at - -
          Rat Genome U34 Set rc_AI232613_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22331803 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline