Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536752_m1
          See other CES5A GE Assays ›
          Gene Symbol:
          Ces5a
          Gene Name:
          carboxylesterase 5A
          Gene Aliases:
          Ces5, Ces7
          Chromosome Location:
          Chr.19: 12097529 - 12136579 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536752_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012056.1 NP_001012056.1 7 - 8 1032 68
          GenBank mRNA AF479659.1 - 7 - 8 1032 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000122.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces5a

          Panther Classification:

          Molecular Function -

          esterase

          Gene Ontology Categories:

          Function(s) Process(es)

          metabolic process
          hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P042840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98213 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950752 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240865 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042840 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P042840 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P042840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P21946 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3281 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89780 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806071 - -
          Affymetrix Rat Gene 1.0 ST Array 10806062 - -
          Affymetrix Rat Gene 1.0 ST Array 10806061 - -
          Affymetrix Rat Gene 1.0 ST Array 10806072 - -
          Affymetrix Rat Gene 1.0 ST Array 10806064 - -
          Affymetrix Rat Gene 1.0 ST Array 10806069 - -
          Affymetrix Rat Gene 1.0 ST Array 10806067 - -
          Affymetrix Rat Gene 1.0 ST Array 10806065 - -
          Affymetrix Rat Gene 1.0 ST Array 10806058 - -
          Affymetrix Rat Gene 1.0 ST Array 10806060 - -
          Affymetrix Rat Gene 1.0 ST Array 10806066 - -
          Affymetrix Rat Gene 1.0 ST Array 10806063 - -
          Affymetrix Rat Gene 1.1 ST Array 10806057 - -
          Rat Genome 230 2.0 Array 1396034_at - -
          Rat Genome 230 2.0 Array 1396034_PM_at - -
          Affymetrix HT Rat Focus Array 1396034_at - -
          Affymetrix HT Rat Focus Array 1396034_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001000.rn.1 TC1900000122.rn.1 RTA TC
          Rat Expression Set 230 1396034_PM_at - -
          Rat Expression Set 230 1396034_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806057 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21527842 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376497 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375569 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline