Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536763_m1
          See other NUP93 GE Assays ›
          Gene Symbol:
          Nup93
          Gene Name:
          nucleoporin 93
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 11159769 - 11263980 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536763_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nup93
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011925.1 NP_001011925.1 20 - 21 2294 62
          XM_008772282.1 XP_008770504.1 18 - 19 4390 62
          GenBank mRNA BC081925.1 - 20 - 21 2294 7113037 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001012.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nup93

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus
          poly(A)+ mRNA export from nucleus
          nuclear pore complex assembly
          structural constituent of nuclear pore

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1027866 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 668210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 482405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 621826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 901173 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 499108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 904219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462839 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143848 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251317 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70831 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33989 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87096 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139144 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60784 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69017 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138499 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35236 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32923 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150447 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53132 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54859 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167665 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44838 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1027866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1027866 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1027866 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1027866 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90864 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34636 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26063 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 101203 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 105040 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87493 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809389 - -
          Affymetrix Rat Gene 1.0 ST Array 10809383 - -
          Affymetrix Rat Gene 1.0 ST Array 10809387 - -
          Affymetrix Rat Gene 1.0 ST Array 10809382 - -
          Affymetrix Rat Gene 1.0 ST Array 10809375 - -
          Affymetrix Rat Gene 1.0 ST Array 10809391 - -
          Affymetrix Rat Gene 1.0 ST Array 10809374 - -
          Affymetrix Rat Gene 1.0 ST Array 10809376 - -
          Affymetrix Rat Gene 1.0 ST Array 10809386 - -
          Affymetrix Rat Gene 1.0 ST Array 10809388 - -
          Affymetrix Rat Gene 1.0 ST Array 10809377 - -
          Affymetrix Rat Gene 1.0 ST Array 10809381 - -
          Affymetrix Rat Gene 1.0 ST Array 10809372 - -
          Affymetrix Rat Gene 1.0 ST Array 10809373 - -
          Affymetrix Rat Gene 1.0 ST Array 10809380 - -
          Affymetrix Rat Gene 1.0 ST Array 10809385 - -
          Affymetrix Rat Gene 1.0 ST Array 10809384 - -
          Affymetrix Rat Gene 1.0 ST Array 10809378 - -
          Affymetrix Rat Gene 1.0 ST Array 10809379 - -
          Affymetrix Rat Gene 1.1 ST Array 10809370 - -
          Rat Genome 230 2.0 Array 1372519_PM_at - -
          Rat Genome 230 2.0 Array 1372519_at - -
          Affymetrix HT Rat Focus Array 1372519_at - -
          Affymetrix HT Rat Focus Array 1372519_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900008158.rn.1 TC1900001012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008156.rn.1 TC1900001012.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900008157.rn.1 TC1900001012.rn.1 RTA TC
          Rat Expression Set 230 1372519_PM_at - -
          Rat Expression Set 230 1372519_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809370 - -
          Rat Genome U34 Set rc_AI169375_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21408843 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376123 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376338 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline