Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536844_m1
          See other TOM1 GE Assays ›
          Gene Symbol:
          Tom1
          Gene Name:
          target of myb1 (chicken)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 14392367 - 14426482 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536844_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tom1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001008365.1 NP_001008366.1 11 - 12 1177 138
          XM_006255145.2 XP_006255207.1 11 - 12 1269 138
          GenBank mRNA BC083873.1 - 11 - 12 1177 7128979 138
          FQ213905.1 - 8 - 9 849 138

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000141.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tom1

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          clathrin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P060579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847306 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 404735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647300 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71482 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89565 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P060579 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P060579 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P380031 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6991 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63070 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806109 - -
          Affymetrix Rat Gene 1.0 ST Array 10806120 - -
          Affymetrix Rat Gene 1.0 ST Array 10806117 - -
          Affymetrix Rat Gene 1.0 ST Array 10806108 - -
          Affymetrix Rat Gene 1.0 ST Array 10806121 - -
          Affymetrix Rat Gene 1.0 ST Array 10806110 - -
          Affymetrix Rat Gene 1.0 ST Array 10806118 - -
          Affymetrix Rat Gene 1.0 ST Array 10806112 - -
          Affymetrix Rat Gene 1.0 ST Array 10806114 - -
          Affymetrix Rat Gene 1.0 ST Array 10806115 - -
          Affymetrix Rat Gene 1.0 ST Array 10806116 - -
          Affymetrix Rat Gene 1.0 ST Array 10806119 - -
          Affymetrix Rat Gene 1.0 ST Array 10806113 - -
          Affymetrix Rat Gene 1.0 ST Array 10806111 - -
          Affymetrix Rat Gene 1.0 ST Array 10806107 - -
          Affymetrix Rat Gene 1.1 ST Array 10806106 - -
          Rat Genome 230 2.0 Array 1384328_at - -
          Rat Genome 230 2.0 Array 1372020_PM_at - -
          Rat Genome 230 2.0 Array 1372020_at - -
          Rat Genome 230 2.0 Array 1384328_PM_at - -
          Affymetrix HT Rat Focus Array 1384328_PM_at - -
          Affymetrix HT Rat Focus Array 1384328_at - -
          Affymetrix HT Rat Focus Array 1372020_at - -
          Affymetrix HT Rat Focus Array 1372020_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001093.rn.1 TC1900000141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001090.rn.1 TC1900000141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001092.rn.1 TC1900000141.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001091.rn.1 TC1900000141.rn.1 RTA TC
          Rat Expression Set 230 1372020_PM_at - -
          Rat Expression Set 230 1372020_at - -
          Rat Expression Set 230 1384328_PM_at - -
          Rat Expression Set 230 1384328_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806106 - -
          Rat Genome U34 Set rc_AA851725_at - -
          Rat Genome U34 Set rc_AA944401_at - -
          Rat Genome U34 Set rc_AA944401_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21621825 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1351051 - -
          RatRef-12 Expression BeadChip ILMN_51393 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline