Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01536890_m1
          See other COQ9 GE Assays ›
          Gene Symbol:
          Coq9
          Gene Name:
          coenzyme Q9
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 10583855 - 10596848 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01536890_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coq9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001035257.1 NP_001030334.1 6 - 7 701 63
          GenBank mRNA BC079370.1 - 6 - 7 687 63
          BC104702.1 - 6 - 7 687 63
          BC127449.1 - 6 - 7 702 8365327 63
          FQ212517.1 - 6 - 7 706 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000988.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coq9

          Gene Ontology Categories:

          Function(s)

          mitochondrial electron transport, NADH to ubiquinone
          ubiquinone biosynthetic process

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P726293 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126252 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388107 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3340 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P726293 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P726293 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P726293 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82401 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15433 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6702 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10809252 - -
          Affymetrix Rat Gene 1.0 ST Array 10809260 - -
          Affymetrix Rat Gene 1.0 ST Array 10809257 - -
          Affymetrix Rat Gene 1.0 ST Array 10809258 - -
          Affymetrix Rat Gene 1.0 ST Array 10809253 - -
          Affymetrix Rat Gene 1.0 ST Array 10809259 - -
          Affymetrix Rat Gene 1.0 ST Array 10809256 - -
          Affymetrix Rat Gene 1.0 ST Array 10809255 - -
          Affymetrix Rat Gene 1.0 ST Array 10809254 - -
          Affymetrix Rat Gene 1.1 ST Array 10809251 - -
          Rat Genome 230 2.0 Array 1388570_at - -
          Rat Genome 230 2.0 Array 1390491_PM_at - -
          Rat Genome 230 2.0 Array 1388570_PM_at - -
          Rat Genome 230 2.0 Array 1390491_at - -
          Affymetrix HT Rat Focus Array 1388570_PM_at - -
          Affymetrix HT Rat Focus Array 1390491_PM_at - -
          Affymetrix HT Rat Focus Array 1388570_at - -
          Affymetrix HT Rat Focus Array 1390491_at - -
          Affymetrix Rat Clariom S Assay PSR1900007890.rn.1 TC1900000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007891.rn.1 TC1900000988.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900007892.rn.1 TC1900000988.rn.1 RTA TC
          Rat Expression Set 230 1388570_PM_at - -
          Rat Expression Set 230 1388570_at - -
          Rat Expression Set 230 1390491_PM_at - -
          Rat Expression Set 230 1390491_at - -
          Affymetrix Rat Gene 1.0 ST Array 10809251 - -
          Rat Genome U34 Set rc_AI232494_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1530525 - -
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370640 - -
          RatRef-12 Expression BeadChip ILMN_161363 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline