Search
Search
| Gene Symbol: Dok4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108438.2 | NP_001101908.1 | 2 - 3 | 352 | 84 | |
| XM_008772314.1 | XP_008770536.1 | 1 - 2 | 293 | 84 | ||
| XM_008772315.1 | XP_008770537.1 | 2 - 3 | 385 | 84 | ||
| XM_008772316.1 | XP_008770538.1 | 2 - 3 | 382 | 84 | ||
| XM_008772317.1 | XP_008770539.1 | 1 - 2 | 292 | 84 | ||
| GenBank mRNA | BC158749.1 | - | 2 - 3 | 352 | 9101502 | 84 |
Set Membership: |
|