Search
Search

RGD1304704| Gene Symbol: RGD1304704 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047874.1 | NP_001041339.1 | 3 - 4 | 289 | 74 | |
| XM_006251636.2 | XP_006251698.1 | 2 - 3 | 295 | 74 | ||
| XM_006251638.2 | XP_006251700.1 | 2 - 3 | 296 | 74 | ||
| GenBank mRNA | AY539943.1 | - | 3 - 4 | 289 | 74 | |
| FQ215371.1 | - | 2 - 3 | 238 | 74 | ||
| FQ220810.1 | - | 2 - 3 | 282 | 74 | ||
| FQ224955.1 | - | 2 - 3 | 289 | 74 | ||
| FQ229372.1 | - | 2 - 3 | 292 | 74 | ||
| FQ229497.1 | - | 2 - 3 | 276 | 74 | ||
Set Membership: |
|