Search
Search
| Gene Symbol: Ctgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022266.2 | NP_071602.1 | 2 - 3 | 503 | 56 | |
| GenBank mRNA | AF120275.1 | - | 2 - 3 | 503 | 56 | |
| BC072503.1 | - | 2 - 3 | 493 | 7098774 | 56 | |
Set Membership: |
|