Search
Search
| Gene Symbol: Ncoa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006227758.2 | XP_006227820.1 | 12 - 13 | 2578 | 60 | |
| XM_006227760.2 | XP_006227822.1 | 2 - 3 | 1697 | 60 | ||
| XM_008758675.1 | XP_008756897.1 | 12 - 13 | 2545 | 60 | ||
Set Membership: |
|