Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01537784_m1
          See other FAM3D GE Assays ›
          Gene Symbol:
          Fam3d
          Gene Name:
          family with sequence similarity 3, member D
          Gene Aliases:
          Oit1
          Chromosome Location:
          Chr.15: 18321757 - 18357252 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01537784_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam3d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106027.1 NP_001099497.1 8 - 9 750 75
          XM_006251752.2 XP_006251814.1 8 - 9 1455 75
          XM_006251753.2 XP_006251815.1 8 - 9 775 75
          XM_008770502.1 XP_008768724.1 3 - 4 1708 75
          XM_008770503.1 XP_008768725.1 6 - 7 1161 75
          XM_008770504.1 XP_008768726.1 7 - 8 879 75
          XM_008770505.1 XP_008768727.1 8 - 9 2383 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000172.rn.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          antimicrobial response protein

          Gene Ontology Categories:

          Function(s)

          negative regulation of insulin secretion

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P011869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710460 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 461250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 863079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 386155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 372364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188812 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268995 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710112 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 261707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135186 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111338 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47580 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37289 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011869 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P011869 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P011869 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P508643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102288 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10779461 - -
          Affymetrix Rat Gene 1.0 ST Array 10779460 - -
          Affymetrix Rat Gene 1.0 ST Array 10779458 - -
          Affymetrix Rat Gene 1.0 ST Array 10779459 - -
          Affymetrix Rat Gene 1.0 ST Array 10779462 - -
          Affymetrix Rat Gene 1.0 ST Array 10779457 - -
          Affymetrix Rat Gene 1.0 ST Array 10779463 - -
          Affymetrix Rat Gene 1.1 ST Array 10779456 - -
          Rat Genome 230 2.0 Array 1383898_at - -
          Rat Genome 230 2.0 Array 1383898_PM_at - -
          Affymetrix HT Rat Focus Array 1383898_PM_at - -
          Affymetrix HT Rat Focus Array 1383898_at - -
          Affymetrix Rat Clariom S Assay PSR1500001282.rn.1 TC1500000172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001285.rn.1 TC1500000172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001281.rn.1 TC1500000172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001284.rn.1 TC1500000172.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500001286.rn.1 TC1500000172.rn.1 RTA TC
          Rat Expression Set 230 1383898_PM_at - -
          Rat Expression Set 230 1383898_at - -
          Affymetrix Rat Gene 1.0 ST Array 10779456 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21315356 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline