Search
Search

Dnase1l3| Gene Symbol: Dnase1l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053907.1 | NP_446359.1 | 4 - 5 | 653 | 123 | |
| GenBank mRNA | AF039852.1 | - | 4 - 5 | 603 | 123 | |
| BC088122.1 | - | 4 - 5 | 651 | 7325326 | 123 | |
| FQ226037.1 | - | 4 - 5 | 480 | 123 | ||
| FQ227612.1 | - | 4 - 5 | 480 | 123 | ||
| FQ230283.1 | - | 4 - 5 | 480 | 123 | ||
| FQ230990.1 | - | 4 - 5 | 480 | 123 | ||
| FQ231646.1 | - | 4 - 5 | 479 | 123 | ||
| FQ231744.1 | - | 4 - 5 | 479 | 123 | ||
| FQ231870.1 | - | 4 - 5 | 483 | 123 | ||
| FQ231939.1 | - | 4 - 5 | 480 | 123 | ||
| FQ231976.1 | - | 4 - 5 | 536 | 123 | ||
| FQ233228.1 | - | 4 - 5 | 536 | 123 | ||
| FQ234709.1 | - | 4 - 5 | 536 | 123 | ||
| U75689.1 | - | 4 - 5 | 653 | 123 | ||
Set Membership: |
|