Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538035_m1
          See other NETO2 GE Assays ›
          Gene Symbol:
          Neto2
          Gene Name:
          neuropilin (NRP) and tolloid (TLL)-like 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 22450579 - 22486530 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538035_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Neto2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001107417.2 NP_001100887.2 2 - 3 232 74
          GenBank mRNA GQ149762.1 - 2 - 3 232 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000254.rn.1

          Set Membership:

          Amplicon length between 71 and 85 Protease 3' Most Recommended Assay Rattus norvegicus Probe spans exons

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of kainate selective glutamate receptor activity
          positive regulation of excitatory postsynaptic potential
          ionotropic glutamate receptor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P036770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 374308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605355 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 940321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361164 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 292250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513189 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42618 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47084 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53465 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 116521 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121009 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71014 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P945840 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1008867 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11415 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44706 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10806441 - -
          Affymetrix Rat Gene 1.0 ST Array 10806442 - -
          Affymetrix Rat Gene 1.0 ST Array 10806444 - -
          Affymetrix Rat Gene 1.0 ST Array 10806438 - -
          Affymetrix Rat Gene 1.0 ST Array 10806443 - -
          Affymetrix Rat Gene 1.0 ST Array 10806439 - -
          Affymetrix Rat Gene 1.0 ST Array 10806440 - -
          Affymetrix Rat Gene 1.1 ST Array 10806436 - -
          Rat Genome 230 2.0 Array 1384904_PM_at - -
          Rat Genome 230 2.0 Array 1393729_PM_at - -
          Rat Genome 230 2.0 Array 1393729_at - -
          Rat Genome 230 2.0 Array 1384904_at - -
          Rat Genome 230 2.0 Array 1392999_PM_at - -
          Rat Genome 230 2.0 Array 1392999_at - -
          Affymetrix HT Rat Focus Array 1384904_at - -
          Affymetrix HT Rat Focus Array 1393729_at - -
          Affymetrix HT Rat Focus Array 1393729_PM_at - -
          Affymetrix HT Rat Focus Array 1392999_PM_at - -
          Affymetrix HT Rat Focus Array 1392999_at - -
          Affymetrix HT Rat Focus Array 1384904_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900001918.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001924.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001919.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001927.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001926.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001920.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001921.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001925.rn.1 TC1900000254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900001928.rn.1 TC1900000254.rn.1 RTA TC
          Rat Expression Set 230 1393729_PM_at - -
          Rat Expression Set 230 1392999_at - -
          Rat Expression Set 230 1384904_PM_at - -
          Rat Expression Set 230 1392999_PM_at - -
          Rat Expression Set 230 1384904_at - -
          Rat Expression Set 230 1393729_at - -
          Affymetrix Rat Gene 1.0 ST Array 10806436 - -
          Rat Genome U34 Set rc_AI059306_at - -
          Rat Genome U34 Set rc_AI144801_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline