Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538561_m1
          See other MAP3K8 GE Assays ›
          Gene Symbol:
          Map3k8
          Gene Name:
          mitogen-activated protein kinase kinase kinase 8
          Gene Aliases:
          D17TPL2, Tpl-2, Tpl2
          Chromosome Location:
          Chr.17: 56109403 - 56129925 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538561_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Map3k8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053847.1 NP_446299.1 7 - 8 1590 72
          XM_006254048.2 XP_006254110.1 7 - 8 1610 72
          GenBank mRNA M94454.1 - 7 - 8 1590 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000804.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Map3k8

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          immune system process
          protein phosphorylation
          apoptotic process
          signal transduction by protein phosphorylation
          intracellular signal transduction
          regulation of cell differentiation
          magnesium ion binding
          receptor signaling protein serine/threonine kinase activity
          MAP kinase kinase kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 922450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195032 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334421 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144962 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15587 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15587 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15587 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15587 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60894 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16855 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795603 - -
          Affymetrix Rat Gene 1.0 ST Array 10795606 - -
          Affymetrix Rat Gene 1.0 ST Array 10795607 - -
          Affymetrix Rat Gene 1.0 ST Array 10795605 - -
          Affymetrix Rat Gene 1.0 ST Array 10795610 - -
          Affymetrix Rat Gene 1.0 ST Array 10795609 - -
          Affymetrix Rat Gene 1.0 ST Array 10795604 - -
          Affymetrix Rat Gene 1.0 ST Array 10795608 - -
          Affymetrix Rat Gene 1.1 ST Array 10795602 - -
          Rat Genome 230 2.0 Array 1369393_PM_at - -
          Rat Genome 230 2.0 Array 1369393_at - -
          Affymetrix HT Rat Focus Array 1369393_PM_at - -
          Affymetrix HT Rat Focus Array 1369393_at - -
          Affymetrix Rat Clariom S Assay PSR1700005390.rn.1 TC1700000804.rn.1 RTA TC
          Rat Expression Set 230 1369393_at - -
          Rat Expression Set 230 1369393_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10795602 - -
          Rat Genome U34 Set M94454_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20903600 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359072 - -
          RatRef-12 Expression BeadChip ILMN_59600 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline