Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01538592_m1
          See other INHBA GE Assays ›
          Gene Symbol:
          Inhba
          Gene Name:
          inhibin beta-A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 51894598 - 51920674 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01538592_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Inhba
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017128.2 NP_058824.1 2 - 3 562 126
          XM_006254001.2 XP_006254063.1 3 - 4 4221 126
          XM_008771712.1 XP_008769934.1 2 - 3 4139 126
          XM_008771713.1 XP_008769935.1 4 - 5 5929 126
          XM_008771714.1 XP_008769936.1 4 - 5 2709 126
          XM_008771715.1 XP_008769937.1 4 - 5 2611 126
          XM_008771716.1 XP_008769938.1 4 - 5 989 126
          GenBank mRNA M37482.1 - 2 - 3 554 126

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001906.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Inhba

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          G1/S transition of mitotic cell cycle
          ovarian follicle development
          mesoderm formation
          hair follicle development
          hematopoietic progenitor cell differentiation
          regulation of transcription from RNA polymerase II promoter
          cell cycle arrest
          negative regulation of cell proliferation
          male gonad development
          positive regulation of pathway-restricted SMAD protein phosphorylation
          negative regulation of cell growth
          positive regulation of cellular protein metabolic process
          activin receptor signaling pathway
          endodermal cell differentiation
          growth
          odontogenesis
          response to drug
          hemoglobin biosynthetic process
          progesterone secretion
          regulation of apoptotic process
          regulation of MAPK cascade
          positive regulation of erythrocyte differentiation
          negative regulation of cell cycle
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of follicle-stimulating hormone secretion
          mesodermal cell differentiation
          cell development
          palate development
          positive regulation of ovulation
          SMAD protein signal transduction
          eyelid development in camera-type eye
          cellular response to follicle-stimulating hormone stimulus
          cellular response to cholesterol
          extrinsic apoptotic signaling pathway
          positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          receptor binding
          cytokine activity
          transforming growth factor beta receptor binding
          hormone activity
          growth factor activity
          peptide hormone binding
          inhibin binding
          identical protein binding
          protein heterodimerization activity
          type II activin receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P030654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412998 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779163 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676721 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51571 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14939 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P030654 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P030654 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P459144 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89016 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 87342 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10798705 - -
          Affymetrix Rat Gene 1.0 ST Array 10798704 - -
          Affymetrix Rat Gene 1.0 ST Array 10798703 - -
          Affymetrix Rat Gene 1.1 ST Array 10798702 - -
          Rat Genome 230 2.0 Array 1369012_PM_at - -
          Rat Genome 230 2.0 Array 1369012_at - -
          Affymetrix HT Rat Focus Array 1369012_PM_at - -
          Affymetrix HT Rat Focus Array 1369012_at - -
          Affymetrix Rat Clariom S Assay PSR1700012862.rn.1 TC1700001906.rn.1 RTA TC
          Rat Expression Set 230 1369012_at - -
          Rat Expression Set 230 1369012_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798702 - -
          Rat Genome U34 Set M37482_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22296458 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370619 - -
          RatRef-12 Expression BeadChip ILMN_61229 - -

          Back To Top
           

          Citations (1)

          Presence, actions, and regulation of myostatin in rat uterus and myometrial cells.
          Authors
          Ciarmela P, Wiater E, Smith SM, Vale W
          Journal
          Endocrinology - Page No 906 (Sun Feb 01 00:00:00 EST 2009)
          Volume (Year)
          150 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline