Search
Search
| Gene Symbol: Add3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164103.1 | NP_001157575.1 | 3 - 4 | 546 | 77 | |
| NM_031552.2 | NP_113740.2 | 3 - 4 | 546 | 77 | ||
| XM_006231599.2 | XP_006231661.1 | 3 - 4 | 399 | 77 | ||
| XM_006231600.2 | XP_006231662.1 | 4 - 5 | 646 | 77 | ||
| XM_006231601.2 | XP_006231663.1 | 3 - 4 | 1245 | 77 | ||
| XM_006231602.2 | XP_006231664.1 | 3 - 4 | 1247 | 77 | ||
| XM_008760510.1 | XP_008758732.1 | 3 - 4 | 389 | 77 | ||
| XM_008760511.1 | XP_008758733.1 | 3 - 4 | 1244 | 77 | ||
| GenBank mRNA | FM072500.1 | - | 3 - 4 | 381 | 77 | |
| U35775.1 | - | 463 | 77 | |||
Set Membership: |
|