Search
Search
| Gene Symbol: Sspo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007016.1 | NP_001007017.1 | 80 - 81 | 12168 | 71 | |
| XM_008762902.1 | XP_008761124.1 | 81 - 82 | 13639 | 71 | ||
| GenBank mRNA | AJ629845.1 | - | 80 - 81 | 12168 | 71 | |
Set Membership: |
|