Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01638246_g1
          See other SNRPN GE Assays ›
          Gene Symbol:
          Snrpn Snurf
          Gene Name:
          small nuclear ribonucleoprotein polypeptide N
          SNRPN upstream reading frame
          Gene Aliases:
          FE 294, FE 294 psi, FE294, FE294 psi
          -
          Chromosome Location:
          Chr.1: 195074328 - 195096460 on Build Rnor_6.0
          Chromosome Location:
          Chr.1: 195074328 - 195096694 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Rn01638246_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Snrpn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031117.2 NP_112379.1 9 - 10 1282 122
          GenBank mRNA BC087671.1 - 5 - 6 686 7316987 122
          M29293.1 - 9 - 10 1282 122
          M29294.1 - 5 - 6 586 122
          Gene Symbol: Snurf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270712.1 NP_001257641.1 10 - 11 1306 122
          NM_130738.2 NP_570094.1 9 - 10 1171 122

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Snrpn
          Click here to find siRNAs for Gene Snurf

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hormone
          molecular_function
          RNA binding
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P038342 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094474 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P013167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 130878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11845 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 82025 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152048 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094474 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P038342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094474 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P038342 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094474 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P157690 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55265 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34097 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10722495 - -
          Affymetrix Rat Gene 1.0 ST Array 10722493 - -
          Affymetrix Rat Gene 1.0 ST Array 10722492 - -
          Affymetrix Rat Gene 1.0 ST Array 10722490 - -
          Affymetrix Rat Gene 1.0 ST Array 10722496 - -
          Affymetrix Rat Gene 1.0 ST Array 10798944 - -
          Affymetrix Rat Gene 1.0 ST Array 10722497 - -
          Affymetrix Rat Gene 1.0 ST Array 10798942 - -
          Affymetrix Rat Gene 1.0 ST Array 10722491 - -
          Affymetrix Rat Gene 1.0 ST Array 10722494 - -
          Affymetrix Rat Gene 1.1 ST Array 10798941 - -
          Affymetrix Rat Gene 1.1 ST Array 10722489 - -
          Affymetrix Rat Gene 1.1 ST Array 10798943 - -
          Rat Genome 230 2.0 Array 1368098_PM_a_at - -
          Rat Genome 230 2.0 Array 1369085_s_at - -
          Rat Genome 230 2.0 Array 1369085_PM_s_at - -
          Rat Genome 230 2.0 Array 1388259_at - -
          Rat Genome 230 2.0 Array 1368098_a_at - -
          Rat Genome 230 2.0 Array 1388259_PM_at - -
          Affymetrix HT Rat Focus Array 1388259_at - -
          Affymetrix HT Rat Focus Array 1368098_PM_a_at - -
          Affymetrix HT Rat Focus Array 1368098_a_at - -
          Affymetrix HT Rat Focus Array 1388259_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100051745.rn.1 TC0100006618.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051749.rn.1 TC0100006618.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051746.rn.1 TC0100006618.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100051747.rn.1 TC0100006618.rn.1 RTA TC
          Rat Expression Set 230 1388259_PM_at - -
          Rat Expression Set 230 1369085_s_at - -
          Rat Expression Set 230 1388259_at - -
          Rat Expression Set 230 1368098_PM_a_at - -
          Rat Expression Set 230 1369085_PM_s_at - -
          Rat Expression Set 230 1368098_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10798941 - -
          Affymetrix Rat Gene 1.0 ST Array 10722489 - -
          Rat Genome U34 Set M29293_at - -
          Rat Genome U34 Set X73411mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22236299 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365045 - -
          RatRef-12 Expression BeadChip ILMN_55162 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline