Search
Search
| Gene Symbol: Tcea2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057098.1 | NP_476439.1 | 8 - 9 | 901 | 72 | |
| XM_006235740.1 | XP_006235802.1 | 8 - 9 | 993 | 72 | ||
| XM_006235741.1 | XP_006235803.1 | 8 - 9 | 978 | 72 | ||
| XM_006235742.2 | XP_006235804.1 | 8 - 9 | 967 | 72 | ||
| GenBank mRNA | D12927.1 | - | 8 - 9 | 901 | 72 | |
Set Membership: |
|