Search
Search
| Gene Symbol: Kcnj15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133321.2 | NP_579855.1 | 2 - 3 | 187 | 109 | |
| XM_006248128.2 | XP_006248190.1 | 3 - 4 | 276 | 109 | ||
| XM_006248129.2 | XP_006248191.1 | 3 - 4 | 413 | 109 | ||
| XM_006248130.2 | XP_006248192.1 | 4 - 5 | 575 | 109 | ||
| XM_006248131.2 | XP_006248193.1 | 3 - 4 | 416 | 109 | ||
| XM_006248132.2 | XP_006248194.1 | 3 - 4 | 367 | 109 | ||
| XM_006248133.2 | XP_006248195.1 | 2 - 3 | 151 | 109 | ||
| XM_006248135.2 | XP_006248197.1 | 2 - 3 | 608 | 109 | ||
| XM_008768550.1 | XP_008766772.1 | 3 - 4 | 384 | 109 | ||
| XM_008768551.1 | XP_008766773.1 | 3 - 4 | 338 | 109 | ||
| XM_008768552.1 | XP_008766774.1 | 3 - 4 | 295 | 109 | ||
| XM_008768553.1 | XP_008766775.1 | 2 - 3 | 2551 | 109 | ||
| GenBank mRNA | AY028455.1 | - | 2 - 3 | 188 | 109 | |
Set Membership: |
|