Search
Search
| Gene Symbol: Ap3m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133593.2 | NP_598277.2 | 5 - 6 | 702 | 66 | |
| XM_006251629.1 | XP_006251691.1 | 5 - 6 | 1229 | 66 | ||
| XM_006251630.2 | XP_006251692.1 | 5 - 6 | 748 | 66 | ||
| GenBank mRNA | BC070925.1 | - | 5 - 6 | 702 | 7096586 | 66 |
| L07073.1 | - | 5 - 6 | 715 | 66 | ||
Set Membership: |
|