Search
Search

Ldhal6b| Gene Symbol: Ldhal6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183334.1 | NP_899163.1 | 1 | 1256 | 106 | |
| GenBank mRNA | AY342428.1 | - | 1 | 1256 | 106 | |
| BC078970.1 | - | 1 | 1237 | 7111795 | 106 | |
Set Membership: |
|