Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01640100_m1
          See other NME8 GE Assays ›
          Gene Symbol:
          Nme8
          Gene Name:
          NME/NM23 family member 8
          Gene Aliases:
          Sptrx2, Txndc3
          Chromosome Location:
          Chr.17: 47301463 - 47368543 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01640100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nme8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198792.2 NP_942087.2 14 - 15 1541 72
          XM_006254088.2 XP_006254150.1 14 - 15 1590 72
          GenBank mRNA AF548544.1 - 12 - 13 1248 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000708.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nme8

          Gene Ontology Categories:

          Function(s) Process(es)

          nucleoside diphosphate phosphorylation
          GTP biosynthetic process
          UTP biosynthetic process
          CTP biosynthetic process
          multicellular organism development
          spermatid development
          cell redox homeostasis
          nucleoside diphosphate kinase activity
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P152124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313100 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 72565 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296969 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 586785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911738 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574793 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 317792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 389256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 124354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 931368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 21725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460291 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88970 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100963 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104889 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75938 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P152124 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P152124 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P498687 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66318 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60716 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 81627 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10795366 - -
          Affymetrix Rat Gene 1.0 ST Array 10795380 - -
          Affymetrix Rat Gene 1.0 ST Array 10795383 - -
          Affymetrix Rat Gene 1.0 ST Array 10795365 - -
          Affymetrix Rat Gene 1.0 ST Array 10795374 - -
          Affymetrix Rat Gene 1.0 ST Array 10795369 - -
          Affymetrix Rat Gene 1.0 ST Array 10795379 - -
          Affymetrix Rat Gene 1.0 ST Array 10795375 - -
          Affymetrix Rat Gene 1.0 ST Array 10795378 - -
          Affymetrix Rat Gene 1.0 ST Array 10795371 - -
          Affymetrix Rat Gene 1.0 ST Array 10795381 - -
          Affymetrix Rat Gene 1.0 ST Array 10795376 - -
          Affymetrix Rat Gene 1.0 ST Array 10795382 - -
          Affymetrix Rat Gene 1.0 ST Array 10795370 - -
          Affymetrix Rat Gene 1.0 ST Array 10795367 - -
          Affymetrix Rat Gene 1.0 ST Array 10795368 - -
          Affymetrix Rat Gene 1.0 ST Array 10795377 - -
          Affymetrix Rat Gene 1.1 ST Array 10795363 - -
          Affymetrix Rat Clariom S Assay PSR1700004813.rn.1 TC1700000708.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004815.rn.1 TC1700000708.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700004812.rn.1 TC1700000708.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10795363 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20915859 - -
          ABI Rat Genome Survey Microarray 21935365 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline