Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01640176_m1
          See other DSG4 GE Assays ›
          Gene Symbol:
          Dsg4
          Gene Name:
          desmoglein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.18: 12056113 - 12092858 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01640176_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dsg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_199490.1 NP_955784.1 12 - 13 1941 121
          GenBank mRNA AY314982.1 - 12 - 13 1941 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1800000099.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dsg4

          Panther Classification:

          Molecular Function -

          cadherin

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          cell adhesion
          homophilic cell adhesion via plasma membrane adhesion molecules
          single organismal cell-cell adhesion
          keratinocyte differentiation
          BMP signaling pathway
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P054227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 218477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 827141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 568465 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87278 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 731066 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21802 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 60454 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46883 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 63150 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054227 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P054227 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P054227 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P259745 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 69825 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84499 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800376 - -
          Affymetrix Rat Gene 1.0 ST Array 10800385 - -
          Affymetrix Rat Gene 1.0 ST Array 10800380 - -
          Affymetrix Rat Gene 1.0 ST Array 10800374 - -
          Affymetrix Rat Gene 1.0 ST Array 10800371 - -
          Affymetrix Rat Gene 1.0 ST Array 10800382 - -
          Affymetrix Rat Gene 1.0 ST Array 10800384 - -
          Affymetrix Rat Gene 1.0 ST Array 10800375 - -
          Affymetrix Rat Gene 1.0 ST Array 10800377 - -
          Affymetrix Rat Gene 1.0 ST Array 10800373 - -
          Affymetrix Rat Gene 1.0 ST Array 10800372 - -
          Affymetrix Rat Gene 1.0 ST Array 10800369 - -
          Affymetrix Rat Gene 1.0 ST Array 10800381 - -
          Affymetrix Rat Gene 1.0 ST Array 10800379 - -
          Affymetrix Rat Gene 1.0 ST Array 10800378 - -
          Affymetrix Rat Gene 1.0 ST Array 10800370 - -
          Affymetrix Rat Gene 1.1 ST Array 10800368 - -
          Affymetrix Rat Clariom S Assay PSR1800000749.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000750.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000751.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000746.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000752.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000745.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000748.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800000747.rn.1 TC1800000099.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10800368 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21868301 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1363146 - -
          RatRef-12 Expression BeadChip ILMN_48246 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline