Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01642020_mH
          See other HMOX2 GE Assays ›
          Gene Symbol:
          Hmox2
          Gene Name:
          heme oxygenase (decycling) 2
          Gene Aliases:
          Ho-2
          Chromosome Location:
          Chr.10: 10990034 - 11035496 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01642020_mH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hmox2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277073.1 NP_001264002.1 4 - 5 841 93
          NM_024387.2 NP_077363.1 4 - 5 885 93
          XM_006245827.2 XP_006245889.1 4 - 5 1063 93
          XM_008767500.1 XP_008765722.1 3 - 4 1792 93
          GenBank mRNA BC062061.1 - 4 - 5 800 5599345 93
          FM110439.1 - 1 - 2 146 93
          FM139649.1 - 1 - 2 307 93
          FQ225862.1 - 4 - 5 839 93
          J05405.1 - 4 - 5 877 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004391.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hmox2

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          heme oxidation
          response to oxidative stress
          heme oxygenase (decyclizing) activity
          protein binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473030 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539061 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 73654 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149195 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12438 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12438 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12438 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12438 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P500907 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92376 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70558 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10740683 - -
          Affymetrix Rat Gene 1.0 ST Array 10740682 - -
          Affymetrix Rat Gene 1.0 ST Array 10740684 - -
          Affymetrix Rat Gene 1.1 ST Array 10740681 - -
          Rat Genome 230 2.0 Array 1384358_at - -
          Rat Genome 230 2.0 Array 1367903_PM_at - -
          Rat Genome 230 2.0 Array 1367903_at - -
          Rat Genome 230 2.0 Array 1384358_PM_at - -
          Affymetrix HT Rat Focus Array 1384358_at - -
          Affymetrix HT Rat Focus Array 1384358_PM_at - -
          Affymetrix HT Rat Focus Array 1367903_PM_at - -
          Affymetrix HT Rat Focus Array 1367903_at - -
          Affymetrix Rat Clariom S Assay PSR1000041106.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041104.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041107.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041113.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041105.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041110.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041108.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041109.rn.1 TC1000004391.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000041122.rn.1 TC1000004391.rn.1 RTA TC
          Rat Expression Set 230 1367903_PM_at - -
          Rat Expression Set 230 1367903_at - -
          Rat Expression Set 230 1384358_PM_at - -
          Rat Expression Set 230 1384358_at - -
          Affymetrix Rat Gene 1.0 ST Array 10740681 - -
          Rat Genome U34 Set U05013_at - -
          Rat Genome U34 Set J05405mRNA_s_at - -
          Rat Genome U34 Set rc_AA955669_at - -
          Rat Genome U34 Set J05405mRNA_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21851284 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373053 - -
          RatRef-12 Expression BeadChip ILMN_53237 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline