Search
Search
| Gene Symbol: Ckm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012530.2 | NP_036662.1 | 4 - 5 | 574 | 61 | |
| GenBank mRNA | DN931989.1 | - | 4 - 5 | 573 | 61 | |
| FQ214594.1 | - | 4 - 5 | 550 | 61 | ||
| FQ214655.1 | - | 4 - 5 | 550 | 61 | ||
| FQ214750.1 | - | 4 - 5 | 550 | 61 | ||
| FQ214825.1 | - | 3 - 4 | 371 | 61 | ||
| FQ214931.1 | - | 4 - 5 | 551 | 61 | ||
| FQ215369.1 | - | 3 - 4 | 448 | 61 | ||
| FQ215410.1 | - | 4 - 5 | 550 | 61 | ||
| FQ215629.1 | - | 2 - 3 | 167 | 61 | ||
| FQ215927.1 | - | 3 - 4 | 403 | 61 | ||
| FQ216321.1 | - | 3 - 4 | 402 | 61 | ||
| FQ216507.1 | - | 4 - 5 | 545 | 61 | ||
| FQ216611.1 | - | 2 - 3 | 167 | 61 | ||
| FQ217421.1 | - | 3 - 4 | 415 | 61 | ||
| FQ224848.1 | - | 3 - 4 | 401 | 61 | ||
| FQ224918.1 | - | 4 - 5 | 551 | 61 | ||
| M10140.1 | - | 4 - 5 | 547 | 61 | ||
| M14864.1 | - | 2 - 3 | 220 | 61 | ||
Set Membership: |
|