Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01645175_m1
          See other HPN GE Assays ›
          Gene Symbol:
          Hpn
          Gene Name:
          hepsin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 89534112 - 89549575 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01645175_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hpn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017112.1 NP_058808.1 8 - 9 986 68
          GenBank mRNA BC072688.1 - 8 - 9 890 7098661 68
          X70900.1 - 8 - 9 986 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005184.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hpn

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          sensory perception of sound
          regulation of cell shape
          positive regulation of gene expression
          negative regulation of alkaline phosphatase activity
          negative regulation of epithelial to mesenchymal transition
          positive regulation of plasminogen activation
          positive regulation of cell growth
          basement membrane disassembly
          cholesterol homeostasis
          negative regulation of apoptotic process
          positive regulation by host of viral transcription
          negative regulation of epithelial cell proliferation
          detection of mechanical stimulus involved in sensory perception of sound
          potassium ion transmembrane transport
          cochlea morphogenesis
          response to thyroid hormone
          pilomotor reflex
          positive regulation of hepatocyte proliferation
          positive regulation of thyroid hormone generation
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          calcium-activated potassium channel activity
          serine-type exopeptidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134748 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P134744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 229601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103140 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713654 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 805915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27697 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235086 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54045 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84436 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134748 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134744 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P134748 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134744 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P134748 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P329357 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75212 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720897 - -
          Affymetrix Rat Gene 1.0 ST Array 10720896 - -
          Affymetrix Rat Gene 1.0 ST Array 10720905 - -
          Affymetrix Rat Gene 1.0 ST Array 10720892 - -
          Affymetrix Rat Gene 1.0 ST Array 10720902 - -
          Affymetrix Rat Gene 1.0 ST Array 10720906 - -
          Affymetrix Rat Gene 1.0 ST Array 10720894 - -
          Affymetrix Rat Gene 1.0 ST Array 10720904 - -
          Affymetrix Rat Gene 1.0 ST Array 10720895 - -
          Affymetrix Rat Gene 1.0 ST Array 10720901 - -
          Affymetrix Rat Gene 1.0 ST Array 10720900 - -
          Affymetrix Rat Gene 1.0 ST Array 10720893 - -
          Affymetrix Rat Gene 1.0 ST Array 10720898 - -
          Affymetrix Rat Gene 1.0 ST Array 10720899 - -
          Affymetrix Rat Gene 1.1 ST Array 10720891 - -
          Rat Genome 230 2.0 Array 1368431_at - -
          Rat Genome 230 2.0 Array 1368431_PM_at - -
          Affymetrix HT Rat Focus Array 1368431_PM_at - -
          Affymetrix HT Rat Focus Array 1368431_at - -
          Affymetrix Rat Clariom S Assay PSR0100041873.rn.1 TC0100005184.rn.1 RTA TC
          Rat Expression Set 230 1368431_PM_at - -
          Rat Expression Set 230 1368431_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720891 - -
          Rat Genome U34 Set X70900_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21390672 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360162 - -
          RatRef-12 Expression BeadChip ILMN_55971 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline