Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01645329_s1
          See other ZFP260 GE Assays ›
          Gene Symbol:
          Zfp260
          Gene Name:
          zinc finger protein 260
          Gene Aliases:
          OZF, POZF-1, Znf146, Znf260
          Chromosome Location:
          Chr.1: 88442011 - 88465110 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01645329_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp260
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017364.4 NP_059060.1 2 817 101
          XM_008759183.1 XP_008757405.1 1184 101
          GenBank mRNA BC099805.1 - 1 664 7937786 101
          U56862.1 - 1 655 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001227.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp260

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          multicellular organism development
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P622195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711742 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P622195 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P622195 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P622195 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P622195 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 27278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10705823 - -
          Affymetrix Rat Gene 1.0 ST Array 10705824 - -
          Affymetrix Rat Gene 1.1 ST Array 10705822 - -
          Rat Genome 230 2.0 Array 1369501_at - -
          Rat Genome 230 2.0 Array 1369501_PM_at - -
          Rat Genome 230 2.0 Array 1372817_PM_at - -
          Rat Genome 230 2.0 Array 1372817_at - -
          Affymetrix HT Rat Focus Array 1369501_PM_at - -
          Affymetrix HT Rat Focus Array 1369501_at - -
          Affymetrix HT Rat Focus Array 1372817_at - -
          Affymetrix HT Rat Focus Array 1372817_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100009481.rn.1 TC0100001227.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100009482.rn.1 TC0100001227.rn.1 RTA TC
          Rat Expression Set 230 1369501_PM_at - -
          Rat Expression Set 230 1372817_PM_at - -
          Rat Expression Set 230 1372817_at - -
          Rat Expression Set 230 1369501_at - -
          Affymetrix Rat Gene 1.0 ST Array 10705822 - -
          Rat Genome U34 Set U56862_at - -
          Rat Genome U34 Set rc_AA801244_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20711852 - -
          ABI Rat Genome Survey Microarray 21064992 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357388 - -
          RatRef-12 Expression BeadChip ILMN_60117 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline