Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01645676_m1
          See other NR5A2 GE Assays ›
          Gene Symbol:
          Nr5a2
          Gene Name:
          nuclear receptor subfamily 5, group A, member 2
          Gene Aliases:
          Ftf, Lrh-1
          Chromosome Location:
          Chr.13: 53750470 - 53870287 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01645676_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr5a2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021742.1 NP_068510.1 7 - 8 1375 79
          XM_006249928.2 XP_006249990.1 6 - 7 1483 79
          XM_006249929.2 XP_006249991.1 5 - 6 1223 79
          GenBank mRNA AB012960.1 - 7 - 8 1375 79
          AB012961.1 - 6 - 7 1312 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001494.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr5a2

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          bile acid metabolic process
          intracellular receptor signaling pathway
          epithelial cell differentiation
          regulation of cell proliferation
          cholesterol homeostasis
          steroid hormone mediated signaling pathway
          positive regulation of viral genome replication
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          pancreas morphogenesis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          phospholipid binding
          zinc ion binding
          lipid binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P035940 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P009798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 763315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377161 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835044 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947880 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 670171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 766562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630632 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 321662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908250 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 816014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581513 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 75782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 603937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 612414 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579639 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 516324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230829 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83398 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29598 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34792 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144934 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50103 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92276 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8212 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49159 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68507 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143160 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71689 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009798 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P035940 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P009798 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P035940 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P009798 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12110 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 31165 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35334 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56827 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77345 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1661 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10768121 - -
          Affymetrix Rat Gene 1.0 ST Array 10768126 - -
          Affymetrix Rat Gene 1.0 ST Array 10768117 - -
          Affymetrix Rat Gene 1.0 ST Array 10768124 - -
          Affymetrix Rat Gene 1.0 ST Array 10768118 - -
          Affymetrix Rat Gene 1.0 ST Array 10768119 - -
          Affymetrix Rat Gene 1.0 ST Array 10768125 - -
          Affymetrix Rat Gene 1.0 ST Array 10768120 - -
          Affymetrix Rat Gene 1.0 ST Array 10768123 - -
          Affymetrix Rat Gene 1.0 ST Array 10768122 - -
          Affymetrix Rat Gene 1.0 ST Array 10768127 - -
          Affymetrix Rat Gene 1.1 ST Array 10768116 - -
          Rat Genome 230 2.0 Array 1387573_PM_a_at - -
          Rat Genome 230 2.0 Array 1387573_a_at - -
          Affymetrix HT Rat Focus Array 1387573_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387573_a_at - -
          Affymetrix Rat Clariom S Assay PSR1300010300.rn.1 TC1300001494.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300010301.rn.1 TC1300001494.rn.1 RTA TC
          Rat Expression Set 230 1387573_a_at - -
          Rat Expression Set 230 1387573_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10768116 - -
          Rat Genome U34 Set AB012960_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372043 - -
          RatRef-12 Expression BeadChip ILMN_66659 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline