Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01648917_m1
          See other PDE2A GE Assays ›
          Gene Symbol:
          Pde2a
          Gene Name:
          phosphodiesterase 2A, cGMP-stimulated
          Gene Aliases:
          Pde2, Pde2a2
          Chromosome Location:
          Chr.1: 166534643 - 166626158 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01648917_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pde2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001143847.1 NP_001137319.1 25 - 26 2358 120
          NM_001270604.1 NP_001257533.1 25 - 26 2308 120
          NM_031079.1 NP_112341.1 24 - 25 2183 120
          GenBank mRNA M94540.1 - 3 - 4 220 120
          U21101.1 - 24 - 25 2183 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002167.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Rattus norvegicus Rat Phosphodiesterase 1700 Inventoried Rat Phosphodiesterase Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pde2a

          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cAMP catabolic process
          protein targeting to mitochondrion
          metabolic process
          positive regulation of gene expression
          cAMP-mediated signaling
          cGMP-mediated signaling
          monocyte differentiation
          negative regulation of cAMP biosynthetic process
          regulation of cGMP metabolic process
          negative regulation of protein import into nucleus, translocation
          cellular response to drug
          cellular response to macrophage colony-stimulating factor stimulus
          negative regulation of vascular permeability
          positive regulation of vascular permeability
          cGMP catabolic process
          positive regulation of inflammatory response
          establishment of endothelial barrier
          cellular response to mechanical stimulus
          cellular response to cGMP
          cellular response to transforming growth factor beta stimulus
          cellular response to granulocyte macrophage colony-stimulating factor stimulus
          cyclic-nucleotide phosphodiesterase activity
          cGMP-stimulated cyclic-nucleotide phosphodiesterase activity
          protein binding
          drug binding
          cAMP binding
          cGMP binding
          TPR domain binding
          GAF domain binding
          protein homodimerization activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 63392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 948066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 76357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534433 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 938557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 5048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 166446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 549824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151921 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 137145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862035 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 338199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 33694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 641233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 752944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 441682 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86319 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83526 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7228 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40950 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94691 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160126 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143913 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12503 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12503 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12503 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P226881 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 98925 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71913 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 73088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77342 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13647 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20522 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88314 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58648 - -
          Agilent Rat cDNA Microarray (G4105A) 701288061 - -
          Agilent Rat cDNA Microarray (G4105A) 701284742 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709205 - -
          Affymetrix Rat Gene 1.0 ST Array 10709226 - -
          Affymetrix Rat Gene 1.0 ST Array 10709211 - -
          Affymetrix Rat Gene 1.0 ST Array 10709233 - -
          Affymetrix Rat Gene 1.0 ST Array 10709222 - -
          Affymetrix Rat Gene 1.0 ST Array 10709203 - -
          Affymetrix Rat Gene 1.0 ST Array 10709227 - -
          Affymetrix Rat Gene 1.0 ST Array 10709202 - -
          Affymetrix Rat Gene 1.0 ST Array 10709212 - -
          Affymetrix Rat Gene 1.0 ST Array 10709225 - -
          Affymetrix Rat Gene 1.0 ST Array 10709223 - -
          Affymetrix Rat Gene 1.0 ST Array 10709206 - -
          Affymetrix Rat Gene 1.0 ST Array 10709224 - -
          Affymetrix Rat Gene 1.0 ST Array 10709230 - -
          Affymetrix Rat Gene 1.0 ST Array 10709213 - -
          Affymetrix Rat Gene 1.0 ST Array 10709210 - -
          Affymetrix Rat Gene 1.0 ST Array 10709208 - -
          Affymetrix Rat Gene 1.0 ST Array 10709214 - -
          Affymetrix Rat Gene 1.0 ST Array 10709220 - -
          Affymetrix Rat Gene 1.0 ST Array 10709231 - -
          Affymetrix Rat Gene 1.0 ST Array 10709215 - -
          Affymetrix Rat Gene 1.0 ST Array 10709209 - -
          Affymetrix Rat Gene 1.0 ST Array 10709229 - -
          Affymetrix Rat Gene 1.0 ST Array 10709228 - -
          Affymetrix Rat Gene 1.0 ST Array 10709232 - -
          Affymetrix Rat Gene 1.0 ST Array 10709207 - -
          Affymetrix Rat Gene 1.0 ST Array 10709221 - -
          Affymetrix Rat Gene 1.0 ST Array 10709216 - -
          Affymetrix Rat Gene 1.0 ST Array 10709219 - -
          Affymetrix Rat Gene 1.0 ST Array 10709234 - -
          Affymetrix Rat Gene 1.0 ST Array 10709217 - -
          Affymetrix Rat Gene 1.0 ST Array 10709218 - -
          Affymetrix Rat Gene 1.1 ST Array 10709200 - -
          Rat Genome 230 2.0 Array 1368089_at - -
          Rat Genome 230 2.0 Array 1368089_PM_at - -
          Affymetrix HT Rat Focus Array 1368089_at - -
          Affymetrix HT Rat Focus Array 1368089_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100016819.rn.1 TC0100002167.rn.1 RTA TC
          Rat Expression Set 230 1368089_PM_at - -
          Rat Expression Set 230 1368089_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709200 - -
          Rat Genome U34 Set U21101_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21835181 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372916 - -
          RatRef-12 Expression BeadChip ILMN_65718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-j77tp:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline