Search
Search

Slc29a1| Gene Symbol: Slc29a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031684.2 | NP_113872.1 | 10 - 11 | 1089 | 69 | |
| XM_006244560.1 | XP_006244622.1 | 10 - 11 | 1120 | 69 | ||
| XM_006244561.2 | XP_006244623.1 | 11 - 12 | 1353 | 69 | ||
| XM_006244562.2 | XP_006244624.1 | 10 - 11 | 1292 | 69 | ||
| XM_006244563.2 | XP_006244625.1 | 10 - 11 | 1115 | 69 | ||
| XM_008766879.1 | XP_008765101.1 | 11 - 12 | 1178 | 69 | ||
| GenBank mRNA | AF015304.1 | - | 7 - 8 | 775 | 69 | |
| BC078789.1 | - | 10 - 11 | 1089 | 7132212 | 69 | |
Set Membership: |
|