Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01729641_m1
          See other PLEKHM1 GE Assays ›
          Gene Symbol:
          Plekhm1
          Gene Name:
          pleckstrin homology domain containing, family M (with RUN domain) member 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 91451890 - 91500675 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01729641_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plekhm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009677.1 NP_001009677.1 11 - 12 3167 77
          XM_006247527.2 XP_006247589.1 11 - 12 3207 77
          XM_008768292.1 XP_008766514.1 10 - 11 2987 77
          GenBank mRNA BC087058.1 - 11 - 12 3167 7124252 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003901.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Plekhm1

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular signal transduction
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P691249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 885301 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 356904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935574 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790967 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 213419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 780142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608874 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 525776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 701427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376439 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972395 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86575 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106036 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36522 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114184 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P691249 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P691249 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P691249 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P436588 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41274 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11201 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748064 - -
          Affymetrix Rat Gene 1.0 ST Array 10748063 - -
          Affymetrix Rat Gene 1.0 ST Array 10748067 - -
          Affymetrix Rat Gene 1.0 ST Array 10748072 - -
          Affymetrix Rat Gene 1.0 ST Array 10748062 - -
          Affymetrix Rat Gene 1.0 ST Array 10748070 - -
          Affymetrix Rat Gene 1.0 ST Array 10748068 - -
          Affymetrix Rat Gene 1.0 ST Array 10748073 - -
          Affymetrix Rat Gene 1.0 ST Array 10748069 - -
          Affymetrix Rat Gene 1.0 ST Array 10748065 - -
          Affymetrix Rat Gene 1.0 ST Array 10748066 - -
          Affymetrix Rat Gene 1.0 ST Array 10748071 - -
          Affymetrix Rat Gene 1.1 ST Array 10748061 - -
          Rat Genome 230 2.0 Array 1380867_PM_a_at - -
          Rat Genome 230 2.0 Array 1380867_a_at - -
          Affymetrix HT Rat Focus Array 1380867_PM_a_at - -
          Affymetrix HT Rat Focus Array 1380867_a_at - -
          Affymetrix Rat Clariom S Assay PSR1000035326.rn.1 TC1000003901.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1000035325.rn.1 TC1000003901.rn.1 RTA TC
          Rat Expression Set 230 1380867_a_at - -
          Rat Expression Set 230 1380867_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748061 - -
          Rat Genome U34 Set rc_AI044862_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21966028 - -
          ABI Rat Genome Survey Microarray 22190650 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360981 - -
          RatRef-12 Expression BeadChip ILMN_49812 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline