Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01746945_m1
          See other RGD1309540 GE Assays ›
          Gene Symbol:
          RGD1309540
          Gene Name:
          similar to hypothetical protein MGC40841; similar to hypothetical protein MGC4707
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 80095040 - 80120088 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01746945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: RGD1309540
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013918.1 NP_001013940.1 301 71
          GenBank mRNA BC087159.1 - 301 7134306 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003655.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene RGD1309540

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P107766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527695 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823463 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965489 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 618977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796801 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92688 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 318493 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 479143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 610378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132651 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165389 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256123 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283483 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 310777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 747017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538895 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 106290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910314 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 179330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 428821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 334178 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12763 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671690 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615728 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 32117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 769215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 710630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781418 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62952 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590400 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140250 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5772 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76990 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 11903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 45697 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 92472 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152270 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 162040 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148671 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114365 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75903 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157946 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161431 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 50612 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122262 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 144501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P107766 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P107766 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P107766 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P107766 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56172 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30143 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35747 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45814 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71104 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26468 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76943 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 45332 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6667 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10847380 - -
          Affymetrix Rat Gene 1.0 ST Array 10847382 - -
          Affymetrix Rat Gene 1.0 ST Array 10847381 - -
          Affymetrix Rat Gene 1.0 ST Array 10847378 - -
          Affymetrix Rat Gene 1.0 ST Array 10847385 - -
          Affymetrix Rat Gene 1.0 ST Array 10847377 - -
          Affymetrix Rat Gene 1.0 ST Array 10847379 - -
          Affymetrix Rat Gene 1.0 ST Array 10847384 - -
          Affymetrix Rat Gene 1.0 ST Array 10847383 - -
          Affymetrix Rat Gene 1.1 ST Array 10847375 - -
          Rat Genome 230 2.0 Array 1372906_PM_at - -
          Rat Genome 230 2.0 Array 1372906_at - -
          Affymetrix HT Rat Focus Array 1372906_PM_at - -
          Affymetrix HT Rat Focus Array 1372906_at - -
          Affymetrix Rat Clariom S Assay PSR0300027777.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027779.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027778.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027780.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027773.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027775.rn.1 TC0300003655.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300027774.rn.1 TC0300003655.rn.1 RTA TC
          Rat Expression Set 230 1372906_at - -
          Rat Expression Set 230 1372906_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10847375 - -
          Rat Genome U34 Set rc_AI008796_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline