Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01746964_m1
          See other METTL15 GE Assays ›
          Gene Symbol:
          Mettl15
          Gene Name:
          methyltransferase like 15
          Gene Aliases:
          RGD1304978
          Chromosome Location:
          Chr.3: 100186106 - 100366148 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01746964_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mettl15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001024981.1 NP_001020152.1 4 - 5 652 55
          XM_006234686.2 XP_006234748.1 5 - 6 824 55
          XM_006234687.2 XP_006234749.1 4 - 5 723 55
          XM_006234688.2 XP_006234750.1 5 - 6 819 55
          XM_006234689.2 XP_006234751.1 5 - 6 798 55
          XM_008762084.1 XP_008760306.1 5 - 6 907 55
          XM_008762085.1 XP_008760307.1 5 - 6 904 55
          XM_008762086.1 XP_008760308.1 5 - 6 886 55
          XM_008762087.1 XP_008760309.1 5 - 6 903 55
          GenBank mRNA BC091215.1 - 4 - 5 652 7383613 55

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003831.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mettl15

          Panther Classification:

          Molecular Function -

          methyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          rRNA base methylation
          rRNA (cytosine-N4-)-methyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P094060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463194 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 403371 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 59166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 906061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66951 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262886 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300040 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699935 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929661 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879332 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160395 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87811 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117072 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 813925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 598509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709810 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854095 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760736 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 712843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 220358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654994 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 308885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 300354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 903609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927119 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851127 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 665795 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 114318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503643 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917359 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46707 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95846 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168947 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140547 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 112942 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115633 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67608 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125085 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99527 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137060 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168590 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094060 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P094060 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P094060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46383 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57158 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56793 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104098 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 102970 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11246 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79324 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4060 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26505 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15561 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10848023 - -
          Affymetrix Rat Gene 1.0 ST Array 10848022 - -
          Affymetrix Rat Gene 1.0 ST Array 10848024 - -
          Affymetrix Rat Gene 1.0 ST Array 10848021 - -
          Affymetrix Rat Gene 1.1 ST Array 10848019 - -
          Affymetrix Rat Clariom S Assay PSR0300029171.rn.1 TC0300003831.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300029172.rn.1 TC0300003831.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10848019 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22106059 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358405 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline