Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747051_m1
          See other PSMD14 GE Assays ›
          Gene Symbol:
          Psmd14
          Gene Name:
          proteasome (prosome, macropain) 26S subunit, non-ATPase, 14
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 47578449 - 47673330 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747051_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmd14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025689.1 NP_001020860.1 11 - 12 1112 69
          GenBank mRNA BC097427.1 - 11 - 12 1112 69
          BC158645.1 - 11 - 12 1060 8363877 69
          FQ228714.1 - 11 - 12 1089 69
          FQ228827.1 - 11 - 12 1087 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000670.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmd14

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          double-strand break repair via nonhomologous end joining
          positive regulation of endopeptidase activity
          response to ethanol
          regulation of proteasomal protein catabolic process
          protein K63-linked deubiquitination
          molecular_function
          endopeptidase activator activity
          proteasome binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P10466 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 314894 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773104 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 23882 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 84079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 205236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 129821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 702680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503062 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437591 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 6809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 801180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 923912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 887570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 395382 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 40627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 178474 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173084 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10466 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P10466 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P10466 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P10466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10845639 - -
          Affymetrix Rat Gene 1.0 ST Array 10845643 - -
          Affymetrix Rat Gene 1.0 ST Array 10845640 - -
          Affymetrix Rat Gene 1.0 ST Array 10845637 - -
          Affymetrix Rat Gene 1.0 ST Array 10845636 - -
          Affymetrix Rat Gene 1.0 ST Array 10845644 - -
          Affymetrix Rat Gene 1.0 ST Array 10845638 - -
          Affymetrix Rat Gene 1.0 ST Array 10845641 - -
          Affymetrix Rat Gene 1.0 ST Array 10845642 - -
          Affymetrix Rat Gene 1.1 ST Array 10845635 - -
          Rat Genome 230 2.0 Array 1375539_PM_at - -
          Rat Genome 230 2.0 Array 1386073_at - -
          Rat Genome 230 2.0 Array 1375539_at - -
          Rat Genome 230 2.0 Array 1383429_at - -
          Rat Genome 230 2.0 Array 1394082_PM_at - -
          Rat Genome 230 2.0 Array 1383429_PM_at - -
          Rat Genome 230 2.0 Array 1386073_PM_at - -
          Rat Genome 230 2.0 Array 1394082_at - -
          Affymetrix HT Rat Focus Array 1383429_at - -
          Affymetrix HT Rat Focus Array 1386073_at - -
          Affymetrix HT Rat Focus Array 1386073_PM_at - -
          Affymetrix HT Rat Focus Array 1394082_PM_at - -
          Affymetrix HT Rat Focus Array 1394082_at - -
          Affymetrix HT Rat Focus Array 1383429_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300005126.rn.1 TC0300000670.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300005124.rn.1 TC0300000670.rn.1 RTA TC
          Rat Expression Set 230 1386073_PM_at - -
          Rat Expression Set 230 1394082_at - -
          Rat Expression Set 230 1383429_PM_at - -
          Rat Expression Set 230 1383429_at - -
          Rat Expression Set 230 1394082_PM_at - -
          Rat Expression Set 230 1386073_at - -
          Affymetrix Rat Gene 1.0 ST Array 10845635 - -
          Rat Genome U34 Set rc_AI043800_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21813105 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline