Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn01747164_m1
          See other SESTD1 GE Assays ›
          Gene Symbol:
          Sestd1
          Gene Name:
          SEC14 and spectrin domains 1
          Gene Aliases:
          RGD1562244
          Chromosome Location:
          Chr.3: 64044111 - 64134977 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn01747164_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sestd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001134514.2 NP_001127986.2 17 - 18 2041 67
          XM_006234402.2 XP_006234464.1 18 - 19 2321 67
          XM_006234404.2 XP_006234466.1 17 - 18 2231 67
          XM_008761892.1 XP_008760114.1 18 - 19 2247 67
          XM_008761893.1 XP_008760115.1 17 - 18 2157 67
          GenBank mRNA BC169110.1 - 17 - 18 2041 7445081 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300003380.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Probe spans exons

          Gene Ontology Categories:

          Process(es)

          phosphatidylserine binding
          1-phosphatidylinositol binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-5-phosphate binding
          phosphatidylcholine binding
          phosphatidylinositol-3-phosphate binding
          phosphatidylinositol-3,4-bisphosphate binding
          phosphatidylinositol-4-phosphate binding
          phosphatidic acid binding
          phosphatidylinositol-3,5-bisphosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P467956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83418 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5403 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467956 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467956 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P467956 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P450214 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P467956 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10846644 - -
          Affymetrix Rat Gene 1.0 ST Array 10846634 - -
          Affymetrix Rat Gene 1.0 ST Array 10846632 - -
          Affymetrix Rat Gene 1.0 ST Array 10846645 - -
          Affymetrix Rat Gene 1.0 ST Array 10846641 - -
          Affymetrix Rat Gene 1.0 ST Array 10846639 - -
          Affymetrix Rat Gene 1.0 ST Array 10846647 - -
          Affymetrix Rat Gene 1.0 ST Array 10846637 - -
          Affymetrix Rat Gene 1.0 ST Array 10846635 - -
          Affymetrix Rat Gene 1.0 ST Array 10846642 - -
          Affymetrix Rat Gene 1.0 ST Array 10846646 - -
          Affymetrix Rat Gene 1.0 ST Array 10846640 - -
          Affymetrix Rat Gene 1.0 ST Array 10846648 - -
          Affymetrix Rat Gene 1.0 ST Array 10846638 - -
          Affymetrix Rat Gene 1.0 ST Array 10846633 - -
          Affymetrix Rat Gene 1.0 ST Array 10846643 - -
          Affymetrix Rat Gene 1.0 ST Array 10846636 - -
          Affymetrix Rat Gene 1.1 ST Array 10846631 - -
          Rat Genome 230 2.0 Array 1388991_PM_at - -
          Rat Genome 230 2.0 Array 1388991_at - -
          Affymetrix HT Rat Focus Array 1388991_at - -
          Affymetrix HT Rat Focus Array 1388991_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300026578.rn.1 TC0300003380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300026579.rn.1 TC0300003380.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300026580.rn.1 TC0300003380.rn.1 RTA TC
          Rat Expression Set 230 1388991_PM_at - -
          Rat Expression Set 230 1388991_at - -
          Affymetrix Rat Gene 1.0 ST Array 10846631 - -
          Rat Genome U34 Set rc_AA946394_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20966039 - -
          ABI Rat Genome Survey Microarray 22097295 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349091 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline